Anti-BAX Mouse Monoclonal Antibody [clone: 2D2]

Proveedor: OriGene
ORIGTA328048EA 509 EUR
ORIGTA328048
Anti-BAX Mouse Monoclonal Antibody [clone: 2D2]
Anticuerpos
Type: Primary
Antigen: BAX
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR