Anti-NEK9 Rabbit Polyclonal Antibody [clone: Poly6241]

Proveedor: OriGene
ORIGTA328043EA 466 EUR
ORIGTA328043
Anti-NEK9 Rabbit Polyclonal Antibody [clone: Poly6241]
Type: Primary
Antigen: NEK9
Clonality: Polyclonal
Clone: Poly6241
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR