Anti-TUBA1C Mouse Monoclonal Antibody [clone: 10D8]

Proveedor: OriGene
ORIGTA328042EA 479 EUR
ORIGTA328042
Anti-TUBA1C Mouse Monoclonal Antibody [clone: 10D8]
Anticuerpos
Type: Primary
Antigen: TUBA1C
Clonality: Monoclonal
Clone: 10D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgM
Reactivity: all species, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR