Anti-PTBP1 Mouse Monoclonal Antibody [clone: 3H7]

Proveedor: OriGene
ORIGTA328040EA 310 EUR
ORIGTA328040
Anti-PTBP1 Mouse Monoclonal Antibody [clone: 3H7]
Anticuerpos
Type: Primary
Antigen: PTBP1
Clonality: Monoclonal
Clone: 3H7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR