Anti-MCL1 Rabbit Polyclonal Antibody [clone: Poly6136]

Proveedor: OriGene
ORIGTA328034EA 466 EUR
ORIGTA328034
Anti-MCL1 Rabbit Polyclonal Antibody [clone: Poly6136]
Type: Primary
Antigen: MCL1
Clonality: Polyclonal
Clone: Poly6136
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR