Anti-PAG1 Rabbit Polyclonal Antibody [clone: Poly6064]

Proveedor: OriGene
ORIGTA328032EA 466 EUR
ORIGTA328032
Anti-PAG1 Rabbit Polyclonal Antibody [clone: Poly6064]
Type: Primary
Antigen: PAG1
Clonality: Polyclonal
Clone: Poly6064
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR