Anti-TPX2 Mouse Monoclonal Antibody [clone: 18D5]

Proveedor: OriGene
ORIGTA328025EA 500 EUR
ORIGTA328025
Anti-TPX2 Mouse Monoclonal Antibody [clone: 18D5]
Anticuerpos
Type: Primary
Antigen: TPX2
Clonality: Monoclonal
Clone: 18D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR