Anti-CHEK2 Mouse Monoclonal Antibody [clone: CHK2909]

Proveedor: OriGene
ORIGTA328023EA 182 EUR
ORIGTA328023
Anti-CHEK2 Mouse Monoclonal Antibody [clone: CHK2909]
Type: Primary
Antigen: CHEK2
Clonality: Monoclonal
Clone: CHK2909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR