Anti-PLK1 Mouse Monoclonal Antibody [clone: 3F8]

Proveedor: OriGene
ORIGTA328019EA 536 EUR
ORIGTA328019
Anti-PLK1 Mouse Monoclonal Antibody [clone: 3F8]
Anticuerpos
Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR