Anti-GAD Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327449EA 524 EUR
ORIGTA327449
Anti-GAD Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: GAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR