Anti-PAX3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327135EA 524 EUR
ORIGTA327135
Anti-PAX3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: PAX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR