Anti-Slc17a8 Mouse Monoclonal Antibody [clone: S34-34]

Proveedor: OriGene
ORIGTA326592EA 623 EUR
ORIGTA326592
Anti-Slc17a8 Mouse Monoclonal Antibody [clone: S34-34]
Anticuerpos
Type: Primary
Antigen: Slc17a8
Clonality: Monoclonal
Clone: S34-34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR