Anti-Slc12a5 Mouse Monoclonal Antibody [clone: S1-12]

Proveedor: OriGene
ORIGTA326588EA 610 EUR
ORIGTA326588
Anti-Slc12a5 Mouse Monoclonal Antibody [clone: S1-12]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:300

Type: Primary
Antigen: Slc12a5
Clonality: Monoclonal
Clone: S1-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR