Anti-Scnn1a Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326579EA 669 EUR
ORIGTA326579
Anti-Scnn1a Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Scnn1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR