Anti-Mapk3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326570EA 544 EUR
ORIGTA326570
Anti-Mapk3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50 Flow Cytometry: 10?g/ml

Type: Primary
Antigen: Mapk3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Bovine, Drosophila, Human, Mouse, Rat, Sheep, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR