Anti-Kcnip1 Mouse Monoclonal Antibody [clone: S55-82]

Proveedor: OriGene
ORIGTA326562EA 623 EUR
ORIGTA326562
Anti-Kcnip1 Mouse Monoclonal Antibody [clone: S55-82]
Anticuerpos
Recommended Dilutions: Immunochemistry: 0.1-1?g/ml

Type: Primary
Antigen: Kcnip1
Clonality: Monoclonal
Clone: S55-82
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR