Anti-Kcnb2 Mouse Monoclonal Antibody [clone: S37-89]

Proveedor: OriGene
ORIGTA326556EA 623 EUR
ORIGTA326556
Anti-Kcnb2 Mouse Monoclonal Antibody [clone: S37-89]
Anticuerpos
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: Kcnb2
Clonality: Monoclonal
Clone: S37-89
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR