Anti-EIF3I Mouse Monoclonal Antibody [clone: 17C11]

Proveedor: OriGene
ORIGTA327980EA 525 EUR
ORIGTA327980
Anti-EIF3I Mouse Monoclonal Antibody [clone: 17C11]
Anticuerpos
Type: Primary
Antigen: EIF3I
Clonality: Monoclonal
Clone: 17C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR