Anti-HIST3H3 Rabbit Polyclonal Antibody [clone: Poly6019]

Proveedor: OriGene
ORIGTA327972EA 466 EUR
ORIGTA327972
Anti-HIST3H3 Rabbit Polyclonal Antibody [clone: Poly6019]
Type: Primary
Antigen: HIST3H3
Clonality: Polyclonal
Clone: Poly6019
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR