Anti-TPT1 Rabbit Polyclonal Antibody [clone: Poly6165]

Proveedor: OriGene
ORIGTA327968EA 451 EUR
ORIGTA327968
Anti-TPT1 Rabbit Polyclonal Antibody [clone: Poly6165]
Type: Primary
Antigen: TPT1
Clonality: Polyclonal
Clone: Poly6165
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR