Anti-TIMP2 Mouse Monoclonal Antibody [clone: F27P3A4]

Proveedor: OriGene
ORIGTA327966EA 536 EUR
ORIGTA327966
Anti-TIMP2 Mouse Monoclonal Antibody [clone: F27P3A4]
Anticuerpos
Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone: F27P3A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken, Human, Mouse, Pig, Pig, Rabbit, swine, zebraFish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR