Anti-TIMP1 Mouse Monoclonal Antibody [clone: F31P2A5]

Proveedor: OriGene
ORIGTA327965EA 500 EUR
ORIGTA327965
Anti-TIMP1 Mouse Monoclonal Antibody [clone: F31P2A5]
Anticuerpos
Type: Primary
Antigen: TIMP1
Clonality: Monoclonal
Clone: F31P2A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR