Anti-RPS27A Mouse Monoclonal Antibody [clone: P4D1]

Proveedor: OriGene
ORIGTA327958EA 564 EUR
ORIGTA327958
Anti-RPS27A Mouse Monoclonal Antibody [clone: P4D1]
Anticuerpos
Type: Primary
Antigen: RPS27A
Clonality: Monoclonal
Clone: P4D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: all species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR