Anti-POU5F1 Rabbit Polyclonal Antibody [clone: Poly6319]

Proveedor: OriGene
ORIGTA327954EA 466 EUR
ORIGTA327954
Anti-POU5F1 Rabbit Polyclonal Antibody [clone: Poly6319]
Type: Primary
Antigen: POU5F1
Clonality: Polyclonal
Clone: Poly6319
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR