Anti-NBN Rabbit Polyclonal Antibody [clone: Poly6138]

Proveedor: OriGene
ORIGTA327951EA 466 EUR
ORIGTA327951
Anti-NBN Rabbit Polyclonal Antibody [clone: Poly6138]
Type: Primary
Antigen: NBN
Clonality: Polyclonal
Clone: Poly6138
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR