Anti-IRF4 Rat Monoclonal Antibody [clone: IRF4.3E4]

Proveedor: OriGene
ORIGTA327948EA 514 EUR
ORIGTA327948
Anti-IRF4 Rat Monoclonal Antibody [clone: IRF4.3E4]
Anticuerpos
Type: Primary
Antigen: IRF4
Clonality: Monoclonal
Clone: IRF4.3E4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR