Anti-LGALS3 Rabbit Polyclonal Antibody [clone: Poly6132]

Proveedor: OriGene
ORIGTA327944EA 466 EUR
ORIGTA327944
Anti-LGALS3 Rabbit Polyclonal Antibody [clone: Poly6132]
Type: Primary
Antigen: LGALS3
Clonality: Polyclonal
Clone: Poly6132
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR