Anti-Kcna4 Mouse Monoclonal Antibody [clone: S13-31]

Proveedor: OriGene
ORIGTA326550EA 678 EUR
ORIGTA326550
Anti-Kcna4 Mouse Monoclonal Antibody [clone: S13-31]
Anticuerpos
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: Kcna4
Clonality: Monoclonal
Clone: S13-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR