Anti-Hcn2 Mouse Monoclonal Antibody [clone: S71-37]

Proveedor: OriGene
ORIGTA326544EA 645 EUR
ORIGTA326544
Anti-Hcn2 Mouse Monoclonal Antibody [clone: S71-37]
Anticuerpos
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: Hcn2
Clonality: Monoclonal
Clone: S71-37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR