Anti-Grm5 Mouse Monoclonal Antibody [clone: S75-33]

Proveedor: OriGene
ORIGTA326542EA 610 EUR
ORIGTA326542
Anti-Grm5 Mouse Monoclonal Antibody [clone: S75-33]
Anticuerpos
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1.0?g/ml

Type: Primary
Antigen: Grm5
Clonality: Monoclonal
Clone: S75-33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR