Anti-Gria1 Mouse Monoclonal Antibody [clone: S355-1]

Proveedor: OriGene
ORIGTA326534EA 645 EUR
ORIGTA326534
Anti-Gria1 Mouse Monoclonal Antibody [clone: S355-1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Gria1
Clonality: Monoclonal
Clone: S355-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR