Anti-Git1 Mouse Monoclonal Antibody [clone: S39B-8]

Proveedor: OriGene
ORIGTA326533EA 692 EUR
ORIGTA326533
Anti-Git1 Mouse Monoclonal Antibody [clone: S39B-8]
Anticuerpos
Type: Primary
Antigen: Git1
Clonality: Monoclonal
Clone: S39B-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR