Anti-Gabrd Mouse Monoclonal Antibody [clone: S151-3]

Proveedor: OriGene
ORIGTA326532EA 623 EUR
ORIGTA326532
Anti-Gabrd Mouse Monoclonal Antibody [clone: S151-3]
Anticuerpos
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: Gabrd
Clonality: Monoclonal
Clone: S151-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR