Anti-Dlgap1 Mouse Monoclonal Antibody [clone: S127-31]

Proveedor: OriGene
ORIGTA326529EA 678 EUR
ORIGTA326529
Anti-Dlgap1 Mouse Monoclonal Antibody [clone: S127-31]
Anticuerpos
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: Dlgap1
Clonality: Monoclonal
Clone: S127-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR