Anti-Aqp4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326513EA 574 EUR
ORIGTA326513
Anti-Aqp4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-4000; Immunofluorescence: 1:400

Type: Primary
Antigen: Aqp4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR