Anti-MEF2A Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327875EA 564 EUR
ORIGTA327875
Anti-MEF2A Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: MEF2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR