Anti-CASP8 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327364EA 564 EUR
ORIGTA327364
Anti-CASP8 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:50-1:100; Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: CASP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR