Anti-HIST2H4A Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327357EA 466 EUR
ORIGTA327357
Anti-HIST2H4A Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000-1:5000; Immunofluorescence: 1:500-1:1000; Immunoprecipitation: 1:200-1:500; CHImmunoprecipitation: 1:50-1:200

Type: Primary
Antigen: HIST2H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR