Anti-Abcc9 Mouse Monoclonal Antibody [clone: S323A-31]

Proveedor: OriGene
ORIGTA326509EA 574 EUR
ORIGTA326509
Anti-Abcc9 Mouse Monoclonal Antibody [clone: S323A-31]
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1.0?g/ml

Type: Primary
Antigen: Abcc9
Clonality: Monoclonal
Clone: S323A-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR