Anti-TCP1 Rat Monoclonal Antibody [clone: 91a]

Proveedor: OriGene
ORIGTA326507EA 603 EUR
ORIGTA326507
Anti-TCP1 Rat Monoclonal Antibody [clone: 91a]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TCP1
Clonality: Monoclonal
Clone: 91a
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: caenorhabditis elegans, Bovine, Dog, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, saccharomyces cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR