Anti-Syt7 Mouse Monoclonal Antibody [clone: S275-14]

Proveedor: OriGene
ORIGTA326504EA 678 EUR
ORIGTA326504
Anti-Syt7 Mouse Monoclonal Antibody [clone: S275-14]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Syt7
Clonality: Monoclonal
Clone: S275-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR