Anti-SYT6 Mouse Monoclonal Antibody [clone: S270-47]

Proveedor: OriGene
ORIGTA326503EA 574 EUR
ORIGTA326503
Anti-SYT6 Mouse Monoclonal Antibody [clone: S270-47]
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SYT6
Clonality: Monoclonal
Clone: S270-47
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR