Anti-SYT5 Mouse Monoclonal Antibody [clone: S276-15]

Proveedor: OriGene
ORIGTA326501EA 574 EUR
ORIGTA326501
Anti-SYT5 Mouse Monoclonal Antibody [clone: S276-15]
Type: Primary
Antigen: SYT5
Clonality: Monoclonal
Clone: S276-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR