Anti-NRAS Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326495EA 564 EUR
ORIGTA326495
Anti-NRAS Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR