Anti-KCNAB1 Mouse Monoclonal Antibody [clone: S9-40]

Proveedor: OriGene
ORIGTA326484EA 574 EUR
ORIGTA326484
Anti-KCNAB1 Mouse Monoclonal Antibody [clone: S9-40]
Anticuerpos
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml

Type: Primary
Antigen: KCNAB1
Clonality: Monoclonal
Clone: S9-40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR