Anti-ING1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326481EA 665 EUR
ORIGTA326481
Anti-ING1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: ING1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR