Anti-HSPA5 Mouse Monoclonal Antibody [clone: 1H11-1H7]

Proveedor: OriGene
ORIGTA326479EA 614 EUR
ORIGTA326479
Anti-HSPA5 Mouse Monoclonal Antibody [clone: 1H11-1H7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HSPA5
Clonality: Monoclonal
Clone: 1H11-1H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR