Anti-HSP90B1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326478EA 587 EUR
ORIGTA326478
Anti-HSP90B1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 1:80

Type: Primary
Antigen: HSP90B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near C-terminus
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR