Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]

Proveedor: OriGene
ORIGTA326476EA 584 EUR
ORIGTA326476
Anti-HIF1A Mouse Monoclonal Antibody [clone: ESEE122]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:1000-1:8000; Immunofluorescence: 8-21?g/ml

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone: ESEE122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR