Anti-DICER1 Mouse Monoclonal Antibody [clone: S167-7]

Proveedor: OriGene
ORIGTA326471EA 574 EUR
ORIGTA326471
Anti-DICER1 Mouse Monoclonal Antibody [clone: S167-7]
Anticuerpos
Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: DICER1
Clonality: Monoclonal
Clone: S167-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR